Kantrowitz Thoracic Forceps, OR Grade, Sklar
Supplier: Sklar
55-2775 55-2795 55-2810
10197-574EA 258.15 USD InStock
10197-574 10197-576 10197-578
Kantrowitz Thoracic Forceps, OR Grade, Sklar
Forceps
Sklar's Kantrowitz Right Angle Thoracic Forceps are a ratcheted, finger ring cardiovascular instrument used to clamp vessels or hold sutures
The right angle curvature of the tips of these forceps allows greater access to hard to reach areas. Kantrowitz thoracic forceps are similar to Mixter and Finochietto thoracic forceps, however Kantrowitz commonly have finer serrations and partially serrated jaws.
Order Now
ORDER
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty