Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IgG1, lambda Isotype Ctrl Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: G0114F7]

Supplier: Biolegend
401907 401908
10761-236EA 93.89 USD InStock
10761-236 10761-238
Anti-IgG1, lambda Isotype Ctrl Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: G0114F7]
Antibodies
PE/Cy7 Rat IgG1, λ Isotype Ctrl [G0114F7]; Isotype: Rat IgG1, λ; Reactivity: Rat; Apps: FC; Size: 100 μg

Each lot of this rat IgG1, λ isotype control antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.

Type: Control
Antigen: IgG1, lambda Isotype Ctrl
Clonality: Monoclonal
Clone: G0114F7
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG1, lambda
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty