Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IL-12/IL-23 Rat Monoclonal Antibody [clone: C17.8]

Supplier: Biolegend
505307 505308
505307-BLEA 136.2 USD InStock
505307-BL 505308-BL
Anti-IL-12/IL-23 Rat Monoclonal Antibody [clone: C17.8]
Antibodies
Ultra-LEAF™ Purified anti-mouse IL-12/IL-23 p40 [C17.8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ELISA, Neut, ICFC, IP, WB; Size: 1 mg

Each lot of this antibody is quality control tested by ELISA assay. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-12/IL-23
Clonality: Monoclonal
Clone: C17.8
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse;IL-12/IL-23 p40 subunit (monomer;homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40)

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty