Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-XCR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ZET]

Supplier: Biolegend
148203 148204
10756-638EA 172.2 USD InStock
10756-638 10756-640
Anti-XCR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ZET]
Antibodies
PE anti-mouse/rat XCR1 [ZET]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse, Rat; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: XCR1
Clonality: Monoclonal
Clone: ZET
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Mouse;Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty