Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Bcl-2 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: BCL/10C4]

Supplier: Biolegend
633511 633512
10019-848EA 126.1 USD InStock
10019-848 10019-850
Anti-Bcl-2 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: BCL/10C4]
Antibodies
PE/Cy7 anti-Bcl-2 [BCL/10C4]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Rat; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining using our nuclear factor staining protocol. For flow cytometric staining, the suggested use of this reagent is 0.125 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Bcl-2
Clonality: Monoclonal
Clone: BCL/10C4
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse;Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty