Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD115 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 9-4D2-1E4]

Supplier: Biolegend
347307 347308
10020-442EA 192.5 USD InStock
10020-442 10020-444
Anti-CD115 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 9-4D2-1E4]
Antibodies
PE/Cy7 anti-human CD115 (CSF-1R) [9-4D2-1E4]; Isotype: Rat IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD115
Clonality: Monoclonal
Clone: 9-4D2-1E4
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty