Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Histone H3 Phospho (Ser28) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: HTA28]

Supplier: Biolegend
641009 641010
10019-852EA 158.7 USD InStock
10019-852 10019-854
Anti-Histone H3 Phospho (Ser28) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: HTA28]
Antibodies
PE anti-Histone H3 Phospho (Ser28) [HTA28]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Histone H3 Phospho (Ser28)
Clonality: Monoclonal
Clone: HTA28
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty