Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Ki-67 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 16A8]

Supplier: Biolegend
652419 652420
10020-544EA 173 USD InStock
10020-544 10020-546
Anti-Ki-67 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 16A8]
Antibodies
Alexa Fluor® 700 anti-mouse Ki-67 [16A8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC, IF; Size: 100 μg

Each lot of this antibody is quality control tested by our Ki-67 staining protocol below. For flow cytometric staining, the suggested use of this reagent is 0.125 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ki-67
Clonality: Monoclonal
Clone: 16A8
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty