Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Perforin Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: dG9]

Supplier: Biolegend
308119 308120
10757-990EA 217.3 USD InStock
10757-990 10021-026
Anti-Perforin Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: dG9]
Antibodies
Brilliant Violet 510™ anti-human Perforin [dG9]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Bovine (Cow, Cattle); Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Perforin
Clonality: Monoclonal
Clone: dG9
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Cross-Reactivity: Bovine (Cow;Cattle)

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty