CytoSelect™ 96-well Phagocytosis Assays, Cell Biolabs
Supplier: Kestrel Engineering Supplies
CBA-220 CBA-222 CBA-224 CBA-224-5
102963-834EA 689.23 USD InStock
102963-834 102963-836 102964-638 102964-640
CytoSelect™ 96-well Phagocytosis Assays, Cell Biolabs
Assays Cellular Assays
Phagocytosis can be assayed in vitro by measuring the engulfment of a substrate by phagocytic cells
Various substrates may be used including red blood cells, E. coli, and Zymosan (yeast) particles. Each CytoSelect™ Phagocytosis Assay quantifies the engulfment of one of these substrates by macrophages or other phagocytes. Quantitation is done in a 96-well plate on a standard colorimetric ELISA plate reader.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty