StemTAG™ Alkaline Phosphatase Assay Kits, Cell Biolabs
Supplier: Kestrel Engineering Supplies
CBA-300 CBA-301 CBA-302
102963-864EA 486.15 USD InStock
102963-864 102963-866 102963-868 102966-504 102966-506 102966-508
StemTAG™ Alkaline Phosphatase Assay Kits, Cell Biolabs
Assays Enzyme Assays
The StemTAG™ Alkaline Phosphatase Assay Kits provide reagents for monitoring alkaline phosphatase activity via either immunocytochemistry staining or 96-well plate quantitation with either colorimetric or fluorescence readout
For best value choose one of our Alkaline Phosphatase Combo Kits that provide reagents for both ICC and 96-well plate detection.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty