Stille Type Gouge, OR Grade, Sklar
Supplier: Sklar
40-6334 40-6335 40-6336
10195-976EA 253.18 USD InStock
10195-976 10195-978 10195-980
Stille Type Gouge, OR Grade, Sklar
Gouges
Stille gouge also known as a chisel, is used in conjuction with a mallet in neurological and orthopedic procedures for cutting and shaping bone.
  • Industry standard
  • Reusable, latex-free
  • Non sterile
The stille gouge has a straight hexagonal handle for grip and the blade profile widens as it radiates from the handle, distrubuting the force. This product is available with either a beveled blade, flat blade, scooped straight blade or a funnel scoop curved blade. The tip of the blade is available in several different diameters.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty