Human Epidermal Keratinocytes, Lifeline Cell Technology
Supplier: LIFELINE CELL TECHNOLOGY MS
FC-0064 FC-0025 FC-0007
102970-718EA 440.46 USD InStock
102970-718 102970-666 102970-960
Human Epidermal Keratinocytes, Lifeline Cell Technology
Cells
Lifeline® Normal Human Epidermal Keratinocytes Neonatal and Adult ensure the highest viability and plating efficiency.
  • Quality tested
  • Ensure optimal serum-free growth
  • Can be grown without serum, phenol red or antimicrobials
Lifeline® Normal Human Epidermal Keratinocytes Neonatal and Adult are cryopreserved as Primary cells to ensure the highest viability and plating efficiency
Lifeline® Epidermal Keratinocytes are quality tested in Lifeline® DermaLife K Medium to ensure optimal serum-free growth over a period of at least 15 population doublings at rates equal to or greater than serum-supplemented medium. Keratinocytes can be grown without serum, phenol red or antimicrobials when cultured in Lifeline® DermaLife medium.
Cells are isolated from neonatal human foreskin and expanded once in culture vessels before cryopreservation.
Información para pedidos: Lifeline® Normal Human Keratinocytes are available as Neonatal Primary Cells, Adult Primary Cells, or 10-Donor Pool. The 10-Donor Pool offers a wide range of donor demographics for research at an economical price. Lifeline® Keratinocytes are 500,000 cells per vial.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty