Airway Epithelial Medium, Lifeline Cell Technology
Supplier: LIFELINE CELL TECHNOLOGY MS
LL-0023 LS-1047 LM-0007
102970-822EA 215.54 USD InStock
102970-822 102970-860 102970-946
Airway Epithelial Medium, Lifeline Cell Technology
Cell Culture Media
BronchiaLife™ Medium is optimized for the serum-free culture of human airway epithelial cells.
  • Optimized for the serum-free culture of human airway epithelial cells
  • Will grow human bronchial/tracheal, lobar, and small airway epithelial cells
  • Maintains excellent cell morphology
  • Contains no antimicrobials and no phenol red
BronchiaLife™ Medium is optimized for the serum-free culture of human airway epithelial cells. BronchiaLife™ will grow human bronchial/tracheal, lobar, and small airway epithelial cells at rates that meet or exceed other commercially available media while maintaining excellent cell morphology. BronchiaLife™ supports the growth of these cells over a period of at least 15 population doublings. BronchiaLife™ contains no antimicrobials and no phenol red.
BronchiaLife™ basal medium requires the appropriate LifeFactors®. BronchiaLife™ Airway Epithelial Medium Complete Kit includes BronchiaLife™ Basal Medium and BronchiaLife™ LifeFactors® Kit.
Información para pedidos: BronchiaLife™ Medium Complete Kit includes BronchiaLife™ Basal Medium and BronchiaLife™ LifeFactors® Kit. BronchiaLife™ Basal Medium requires the associated LifeFactors®.
BronchiaLife™ LifeFactors® Kit includes the associated supplements and growth factors for use with human airway epithelial cells and BronchiaLife™ Basal Medium. The Kit includes: HLL Supplement 1.25 mL, L-Glutamine 15 mL, Extract P™ 2 mL, TM1 5 mL, Gentamicin and Amphotericin B 0.5 mL.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty