ASI Toxoplasma, Arlington Scientific
Supplier: ARLINGTON SCIENTIFIC INC.
810096AG 810096AM
10853-078KT 455.72 USD InStock
10853-078 10853-080
ASI Toxoplasma, Arlington Scientific
Assays ELISAs
ELISA Kit detects human antibodies to Toxoplasma gondii.
  • Qualitative and semiquantitative detection
  • Detects human IGG or IGM antibodies
ELISA for the qualitative and semiquantitative detection of human IGG/IGM antibodies to Toxoplasma gondii.
Envase: Kit of 96
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty