Quick Release Clamp, Thermo Scientific
Supplier: Epredia
556000 715200
89238-690EA 1259.65 USD InStock
89238-690 89238-734 89238-820
Quick Release Clamp, Thermo Scientific
Clamps General Purpose Clamps
Fix cassettes or embedding rings both longitudinally and horizontally, with orientation on X-, Y-, and Z-axes using this quick-release clamp.
  • Universal Cassette Clamps for all types of commercially available cassettes
  • Quick Release Clamp for embedding rings
  • Zero Orientation Device positions specimen holder flat against the knife or blade (aligns X, Y axis)
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty