Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Standard Ferrules, Fit 0.28 mm int.Ø MXT® Columns, Restek

Supplier: Restek
27066 20232
10852-536PK 60.62 USD InStock
10852-536 10852-564
Standard Ferrules, Fit 0.28 mm int.Ø MXT® Columns, Restek
Fittings and Connectors
Standard ferrules fit 0.28 mm ID MXT® columns.

  • Seal with minimal torque
  • Reusable
  • Preferred for vacuum and high-pressure areas
  • Stable to 400°C

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty