Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Azelaic acid ≥90% (by GC), Technical Grade for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Warning

M-Clarity™ quality level = MQ200

8.20116.0100 8.20116.1000 8.20116.9025 8.20116.9050
EM8.20116.0100CS 3484 USD InStock
EM8.20116.0100 EM8.20116.1000 EM8.20116.9025 EM8.20116.9050
Azelaic acid ≥90% (by GC), Technical Grade for synthesis, Sigma-Aldrich®
Azelaic acid


Formula: C₉H₁₆O₄
Boiling Pt: 371 °C (1013 hPa)
Density: 1.0287 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004432
CAS Number: 123-99-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty