Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-1H-pyrazole-5-boronic acid pinacol ester

Supplier: MATRIX SCIENTIFIC MS

034055-5G 034055-25G
102551-160EA 36.92 USD InStock
102551-160 102551-162
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester
Hazard: Irritant


Formula: C₁₀H₁₇BN₂O₂
Melting Pt: 58…61 °C
MDL Number: MFCD05861380
CAS Number: 847818-74-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty