Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl lactate

Supplier: MATRIX SCIENTIFIC MS

058299-5G 058299-25G 058299-100G
102833-252EA 15.38 USD InStock
102833-252 102833-254 102833-256
Ethyl lactate
Ethyl lactate
Hazard: Flammable


Formula: C₅H₁₀O₃
Boiling Pt: 152-155°C
Density: 1.036
Flash Pt: 46°C
MDL Number: MFCD00065359
CAS Number: 687-47-8
UN: 1192
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty