Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethyl-1-adamantanamine hydrochloride

Supplier: MATRIX SCIENTIFIC MS

060562-5G 060562-25G
102833-300EA 36.92 USD InStock
102833-300 102833-302
3,5-Dimethyl-1-adamantanamine hydrochloride
3,5-Dimethyl-1-adamantanamine hydrochloride
Hazard: Irritant


Formula: C₁₂H₂₁N
MDL Number: MFCD00214336
CAS Number: 41100-52-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty