Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Hydroxyethyl)-2-pyrrolidone

Supplier: MATRIX SCIENTIFIC MS

064284-25G 064284-100G
102833-360EA 106.15 USD InStock
102833-360 102833-362
1-(2-Hydroxyethyl)-2-pyrrolidone
1-(2-Hydroxyethyl)-2-pyrrolidone
TCA listed: T
Hazard: Irritant


Formula: C₆H₁₁NO₂
Boiling Pt: 295 °C
Melting Pt: 20 °C
Density: 1.1435/20°C
MDL Number: MFCD00014103
CAS Number: 3445-11-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty