Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-6-methylpyrimidine

Supplier: MATRIX SCIENTIFIC MS

064531-1G 064531-5G 064531-10G
102663-816EA 64.62 USD InStock
102663-816 102663-818 102663-820
4-Hydroxy-6-methylpyrimidine
4-Hydroxy-6-methylpyrimidine
Hazard: Irritant


Formula: C₅H₆N₂O
Melting Pt: 145…150 °C
MDL Number: MFCD00053575
CAS Number: 3524-87-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty