Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2,4-dichloropyrimidine-5-carboxylate ≥97%

Supplier: MATRIX SCIENTIFIC MS

071103-5G 071103-25G
102519-474EA 144.62 USD InStock
102519-474 102663-862
Ethyl-2,4-dichloropyrimidine-5-carboxylate ≥97%
Ethyl-2,4-dichloropyrimidine-5-carboxylate
Hazard: Irritant-Harmful/Moisture Sensitive/Refrigerate


Formula: C₇H₆Cl₂N₂O₂
MW: 221.04 g/mol
Melting Pt: 36 °C
MDL Number: MFCD09910281
CAS Number: 51940-64-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty