Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-{1H-pyrrolo[2,3-b]pyridin-3-ylmethyl}piperazine-1-carboxylate ≥97%

Supplier: MATRIX SCIENTIFIC MS

071378-500MG 071378-1G
102519-954EA 403.08 USD InStock
102519-954 102519-956
tert-Butyl-4-{1H-pyrrolo[2,3-b]pyridin-3-ylmethyl}piperazine-1-carboxylate ≥97%
tert-Butyl-4-{1H-pyrrolo[2,3-b]pyridin-3-ylmethyl}piperazine-1-carboxylate
Hazard: Irritant


Formula: C₁₇H₂₄N₄O₂
Melting Pt: 187…189 °C
MDL Number: MFCD11054205
CAS Number: 1065100-86-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty