Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Thiophen-2-yl)benzoic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

071835-1G 071835-5G 071835-10G
102521-612EA 213.85 USD InStock
102521-612 102521-614 102521-616
4-(Thiophen-2-yl)benzoic acid ≥95%
4-(Thiophen-2-yl)benzoic acid
Hazard: Irritant


Formula: C₁₁H₈O₂S
MDL Number: MFCD00690742
CAS Number: 29886-62-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty