Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-(4-(Bis(4-fluorophenyl)methyl)piperazin-1-yl)phenyl)methanol ≥95%

Supplier: MATRIX SCIENTIFIC MS

071880-1G 071880-5G 071880-10G
102521-872EA 213.85 USD InStock
102521-872 102521-874 102521-876
(2-(4-(Bis(4-fluorophenyl)methyl)piperazin-1-yl)phenyl)methanol ≥95%
(2-(4-(Bis(4-fluorophenyl)methyl)piperazin-1-yl)phenyl)methanol
Hazard: Irritant


Formula: C₂₄H₂₄F₂N₂O
MW: 394.4600 g/mol
MDL Number: MFCD06797786
CAS Number: 914349-61-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty