Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Phenylcycloheptanamine hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

071976-1G 071976-5G 071976-10G
102522-418EA 710.77 USD InStock
102522-418 102522-420 102522-422
1-Phenylcycloheptanamine hydrochloride ≥95%
1-Phenylcycloheptanamine hydrochloride
Hazard: Irritant


Formula: C₁₃H₂₀ClN
CAS Number: 125802-37-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty