Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Homoserine ≥95%

Supplier: MATRIX SCIENTIFIC MS

072208-5G 072208-25G
102523-662EA 53.85 USD InStock
102523-662 102833-528
L-Homoserine ≥95%
L-Homoserine
Hazard: Irritant


Formula: C₄H₉NO₃
MW: 119.12 g/mol
Melting Pt: 203
MDL Number: MFCD00063090
CAS Number: 672-15-1
Merck Index: 13,04760
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty