Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3R,4R)-3-Amino-4-hydroxypentanoic acid hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

072209-250MG 072209-1G 072209-5G
102523-666EA 400 USD InStock
102523-666 102523-668 102523-670
(3R,4R)-3-Amino-4-hydroxypentanoic acid hydrochloride ≥95%
(3R,4R)-3-Amino-4-hydroxypentanoic acid hydrochloride
Hazard: Irritant


Formula: C₅H₁₂ClNO₃
MDL Number: MFCD01862874
CAS Number: 336182-14-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty