Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1H-Benzo[d]imidazol-2-yl)-4-bromophenol ≥95%

Supplier: MATRIX SCIENTIFIC MS

072354-250MG 072354-1G 072354-5G
102524-408EA 355.38 USD InStock
102524-408 102524-410 102524-412
2-(1H-Benzo[d]imidazol-2-yl)-4-bromophenol ≥95%
2-(1H-Benzo[d]imidazol-2-yl)-4-bromophenol
Hazard: Irritant


Formula: C₁₃H₉BrN₂O
MW: 289.13 g/mol
MDL Number: MFCD00795713
CAS Number: 62871-28-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty