Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-methoxybenzaldehyde ≥95%

Supplier: MATRIX SCIENTIFIC MS

072500-250MG 072500-1G 072500-5G
102525-170EA 427.69 USD InStock
102525-170 102525-172 102525-174
2-Amino-3-methoxybenzaldehyde ≥95%
2-Amino-3-methoxybenzaldehyde
Hazard: Irritant


Formula: C₈H₉NO₂
MDL Number: MFCD09999245
CAS Number: 70127-96-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty