Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methylpyridin-4-ol ≥95%

Supplier: MATRIX SCIENTIFIC MS

072531-1G 072531-5G
102525-346EA 240 USD InStock
102525-346 102525-348
3-Methylpyridin-4-ol ≥95%
3-Methylpyridin-4-ol
Hazard: Irritant


Formula: C₆H₇NO
MW: 109.13
MDL Number: MFCD04114239
CAS Number: 22280-02-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty