Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-m-anisidine ≥95%

Supplier: MATRIX SCIENTIFIC MS

072566-1G 072566-5G 072566-10G
102525-546EA 78.46 USD InStock
102525-546 102525-548 102525-550
2-Chloro-m-anisidine ≥95%
2-Chloro-m-anisidine
Hazard: Toxic


Formula: C₇H₈ClNO
Melting Pt: 50°C
MDL Number: MFCD08691737
CAS Number: 113206-03-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty