Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Oxazol-5-ylmethanamine hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

072644-250MG 072644-1G 072644-5G
102525-970EA 595.38 USD InStock
102525-970 102525-972 102525-974
Oxazol-5-ylmethanamine hydrochloride ≥95%
Oxazol-5-ylmethanamine hydrochloride
Hazard: Irritant


Formula: C₄H₇ClN₂O
MW: 134.56 g/mol
MDL Number: MFCD06804578
CAS Number: 1196156-45-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty