Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoro-4-methoxyaniline ≥95%

Supplier: MATRIX SCIENTIFIC MS

072784-1G 072784-5G 072784-10G
102526-726EA 138.46 USD InStock
102526-726 102526-728 102526-730
2,6-Difluoro-4-methoxyaniline ≥95%
2,6-Difluoro-4-methoxyaniline
Hazard: Irritant


Formula: C₇H₇F₂NO
MW: 159.14
MDL Number: MFCD04115922
CAS Number: 151414-47-0
UN: 2811
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty