Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-nitrophenol ≥95%

Supplier: MATRIX SCIENTIFIC MS

072903-5G 072903-100G 072903-25G
102527-360EA 47.69 USD InStock
102527-360 102527-364 102663-912
2-Amino-3-nitrophenol ≥95%
2-Amino-3-nitrophenol
Hazard: Irritant


Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
MDL Number: MFCD00010875
CAS Number: 603-85-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty