Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-4-pyridineboronic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

072944-1G 072944-5G
102527-576EA 283.08 USD InStock
102527-576 102527-578
2-Methoxy-4-pyridineboronic acid ≥95%
2-Methoxy-4-pyridineboronic acid
Hazard: Irritant/Refrigerate


Formula: C₆H₈BNO₃
MW: 152.9436 g/mol
MDL Number: MFCD07368877
CAS Number: 762262-09-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty