Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Benzenesulfonylbenzoic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

072986-250MG 072986-1G
102527-786EA 569.23 USD InStock
102527-786 102527-788
4-Benzenesulfonylbenzoic acid ≥95%
4-Benzenesulfonylbenzoic acid
Hazard: Irritant


Formula: C₁₃H₁₀O₄S
MW: 262.29 g/mol
MDL Number: MFCD00568775
CAS Number: 5361-54-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty