Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(6-Chloropyrazin-2-yl)acetic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

072997-250MG 072997-1G 072997-5G
102527-840EA 292.31 USD InStock
102527-840 102527-842 102527-844
2-(6-Chloropyrazin-2-yl)acetic acid ≥95%
2-(6-Chloropyrazin-2-yl)acetic acid
Hazard: Irritant


Formula: C₆H₅ClN₂O₂
MW: 172.570 g/mol
MDL Number: MFCD09907855
CAS Number: 930798-25-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty