Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methyl-4-nitro-3-isoxazolecarboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

073002-250MG 073002-1G 073002-5G
102527-866EA 498.46 USD InStock
102527-866 102527-868 102527-870
5-Methyl-4-nitro-3-isoxazolecarboxylic acid ≥95%
5-Methyl-4-nitro-3-isoxazolecarboxylic acid
Hazard: Irritant


Formula: C5H4N2O5
MDL Number: MFCD11520861
CAS Number: 960225-75-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty