Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-nitroanisole ≥95%

Supplier: MATRIX SCIENTIFIC MS

073101-5G 073101-25G 073101-100G
102528-380EA 43.08 USD InStock
102528-380 102663-928 102663-930
2-Fluoro-5-nitroanisole ≥95%
2-Fluoro-5-nitroanisole
Hazard: Irritant


Formula: C₇H₆FNO₃
Melting Pt: 69…71 °C
MDL Number: MFCD07782081
CAS Number: 454-16-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty