Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopropanecarboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

073415-100G 073415-500G
102530-048EA 52.31 USD InStock
102530-048 102551-840
Cyclopropanecarboxylic acid ≥95%
Cyclopropanecarboxylic acid
Hazard: Corrosive


Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 182
Melting Pt: 5 °C
Density: 1.086
Flash Pt: 71 °C
MDL Number: MFCD00001287
CAS Number: 1759-53-1
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty