Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-amino-1-(4-chlorobenzyl)piperidine-4-carboxylate dihydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

073545-250MG 073545-1G 073545-5G
102530-732EA 456.92 USD InStock
102530-732 102530-734 102530-736
Methyl-4-amino-1-(4-chlorobenzyl)piperidine-4-carboxylate dihydrochloride ≥95%
Methyl-4-amino-1-(4-chlorobenzyl)piperidine-4-carboxylate dihydrochloride
Hazard: Irritant


Formula: C₁₄H₂₁Cl₃N₂O₂
MDL Number: MFCD08543515
CAS Number: 1210494-26-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty