Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methylpyrimidine ≥95%

Supplier: MATRIX SCIENTIFIC MS

073690-1G 073690-5G 073690-25G
102531-502EA 75.38 USD InStock
102531-502 102531-504 102663-956
5-Methylpyrimidine ≥95%
5-Methylpyrimidine
Hazard: Flammable Solid


Formula: C₅H₆N₂
Boiling Pt: 152…154 °C
Melting Pt: 33…37 °C
Flash Pt: 53 °C
MDL Number: MFCD00047386
CAS Number: 2036-41-1
UN: 1325
ADR: 4.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty