Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Chloro[1,2,4]triazolo[1,5-a]pyrazine ≥95%

Supplier: MATRIX SCIENTIFIC MS

073738-250MG 073738-1G
102531-746EA 826.15 USD InStock
102531-746 102531-748
8-Chloro[1,2,4]triazolo[1,5-a]pyrazine ≥95%
8-Chloro[1,2,4]triazolo[1,5-a]pyrazine
Hazard: Irritant


Formula: C₅H₃ClN₄
MDL Number: MFCD09834989
CAS Number: 74803-32-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty