Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzo[d]isoxazole-3-carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

073778-250MG 073778-1G 073778-5G
102531-952EA 90.77 USD InStock
102531-952 102531-954 102531-956
Benzo[d]isoxazole-3-carboxylic acid ≥95%
Benzo[d]isoxazole-3-carboxylic acid
Hazard: Irritant


Formula: C₈H₅NO₃
Boiling Pt: 379°C
Density: 1.45
Flash Pt: 183°C
MDL Number: MFCD06659786
CAS Number: 28691-47-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty