Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chlorobenzofuro[3,2-d]pyrimidine ≥95%

Supplier: MATRIX SCIENTIFIC MS

073858-250MG 073858-1G 073858-5G
102533-374EA 498.46 USD InStock
102533-374 102533-376 102533-378
4-Chlorobenzofuro[3,2-d]pyrimidine ≥95%
4-Chlorobenzofuro[3,2-d]pyrimidine
Hazard: Irritant


Formula: C₁₀H₅ClN₂O
MDL Number: MFCD00763943
CAS Number: 39876-88-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty