Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

073888-1G 073888-5G 073888-10G
102533-536EA 427.69 USD InStock
102533-536 102533-538 102533-540
Ethyl-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate ≥95%
Ethyl-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate
Hazard: Irritant


Formula: C₁₀H₁₃NO₃
MDL Number: MFCD00030352
CAS Number: 2199-59-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty