Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Pyrimidineacetic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

073955-250MG 073955-1G 073955-5G
102533-884EA 98.46 USD InStock
102533-884 102533-886 102533-888
5-Pyrimidineacetic acid ≥95%
5-Pyrimidineacetic acid
Hazard: Irritant


Formula: C6H6N2O2
Melting Pt: 172…173 °C
MDL Number: MFCD09608122
CAS Number: 5267-07-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty